• USA Home
  • I2006 - Indole-3-carboxylic acid

EMAIL THIS PAGE TO A FRIEND
I2006 Sigma-Aldrich

Indole-3-carboxylic acid

Synonym: β-Indolylcarboxylic acid, 3-Carboxyindole, 3-Indole formic acid, 3-Indolylcarboxylic acid, Indole-β-carboxylic acid

  • CAS Number 771-50-6

  • Empirical Formula (Hill Notation) C9H7NO2

  • Molecular Weight 161.16

  •  Beilstein/REAXYS Number 129435

  •  EC Number 212-231-6

  •  MDL number MFCD00005624

Purchase

Properties

mp   232-234 °C (dec.) (lit.)
SMILES string   OC(=O)c1c[nH]c2ccccc12
InChI   1S/C9H7NO2/c11-9(12)7-5-10-8-4-2-1-3-6(7)8/h1-5,10H,(H,11,12)
InChI key   KMAKOBLIOCQGJP-UHFFFAOYSA-N

Description

Application

Reactant for preparation of:
• Anticancer agents
• Derivatives of amino acids and peptides
• Serotonin 5-HT4 receptor antagonists
• Primary acylureas
• Inhibitors of Gli1-mediated transcription in the Hedgehog pathway
• Serotonin 5-HT6 antagonists
• Very Late Antigen-4 (VLA-4) sntagonists
• EphB3 receptor tyrosine kinase inhibitors
• Potential Therapeutic Agent for Alzheimer′s Disease
• Vinyl ester pseudotripeptide proteasome inhibitors

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

284734 Indole-3-carboxylic acid, ReagentPlus®, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?