• USA Home
  • I2767 - Insulin sodium salt human

EMAIL THIS PAGE TO A FRIEND
I2767 Sigma-Aldrich

Insulin sodium salt human

recombinant, expressed in E. coli, crystalline, suitable for cell culture

Purchase

Properties

recombinant   expressed in E. coli
assay   ≥90% (HPLC)
form   crystalline
application(s)   cell culture | mammalian: suitable
solubility   10 mM HCl: 20 mg/mL
cation traces   Zn: ≤0.01%
storage temp.   −20°C

Description

Biochem/physiol Actions

Two-chain polypeptide hormone produced by the β-cells of pancreatic islets. Its molecular weight is ~5800 Da. The α and β chains are joined by two interchain disulfide bonds. The α chain contains an intrachain disulfide bond. Insulin regulates the cellular uptake, utilization, and storage of glucose, amino acids, and fatty acids and inhibits the breakdown of glycogen, protein, and fat.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg
RTECS 
NM8900250

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

I2643 Insulin human, recombinant, expressed in yeast (proprietary host)

related product

Product #

Description

Add to Cart

I9278 Insulin solution human, sterile-filtered, BioXtra, suitable for cell culture

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?