• USA Home
  • I3783 - Monoclonal Anti-Integrin αV antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
I3783 Sigma-Aldrich

Monoclonal Anti-Integrin αV antibody produced in mouse

1 mg/mL, clone P2W7, tissue culture supernatant

Synonym: Anti-CD51

Purchase

Properties

Related Categories Alphabetical Index, Angiogenesis, Antibodies, Antibodies for Cell Biology, Antibodies for Hematopoietic Stem Cells,
conjugate   unconjugated
clone   P2W7, monoclonal
biological source   mouse
application(s)   flow cytometry: suitable
species reactivity   human
mol wt   antigen 160 kDa
shipped in   wet ice
storage temp.   2-8°C
antibody form   tissue culture supernatant
isotype   IgG1κ
antibody product type   primary antibodies
concentration   1 mg/mL
UniProt accession no.   P06756
Gene Information   human ... ITGAV(3685)

Description

Immunogen

ocular melanoma cell line expressing high levels of human integrin αV, β2.

Physical form

Solution in 50 mM sodium phosphate, pH 7.5, 100 mM potassium chloride, 150 mM sodium chloride, and 0.5 mg/ml gentamicin sulfate.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?