• USA Home
  • I3903 - Monoclonal Anti-Interleukin-6 antibody produced in mouse

I3903 Sigma

Monoclonal Anti-Interleukin-6 antibody produced in mouse

clone 53325.111, purified immunoglobulin, lyophilized powder

Synonym: Anti-IL-6



Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies to Cytokines and Growth Factors, Antibodies to Interleukins (IL),
biological source   mouse
antibody form   purified immunoglobulin
clone   53325.111, monoclonal
form   lyophilized powder
species reactivity   rat
application(s)   capture ELISA: suitable using a recombinant rat IL-6
isotype   IgG2b
conjugate   unconjugated
storage temp.   −20°C



recombinant rat IL-6 expressed in Escherichia coli.

Preparation Note

Purified using protein G.

Physical form

Lyophilized from 0.2 μm filtered solution in phosphate buffered saline.

Price and Availability

Antibody Explorer

Explore with Confidence

Milli-Q® Water Purification Solutions
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles


Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers


Related Products

comparable product

Product #


Add to Cart

I7901 Monoclonal Anti-Interleukin-6 antibody produced in mouse, clone 6708.11, purified immunoglobulin, lyophilized powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?