• USA Home
  • I5407 - Anti-Interleukin-21 Receptor, Extracellular Domain antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
I5407 Sigma-Aldrich

Anti-Interleukin-21 Receptor, Extracellular Domain antibody produced in rabbit

~0.5 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym: Anti-IL-21R

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies to Cytokines and Growth Factors, Antibodies to Interleukins (IL),
conjugate   unconjugated
clone   polyclonal
biological source   rabbit
application(s)   western blot: 0.5-1 μg/mL using human Raji and A431 cell lysates
species reactivity   human
form   buffered aqueous solution
shipped in   dry ice
storage temp.   −20°C
antibody form   affinity isolated antibody
antibody product type   primary antibodies
concentration   ~0.5 mg/mL
UniProt accession no.   Q9HBE5
Gene Information   human ... IL21R(50615)

Description

Immunogen

synthetic peptide corresponding to amino acids 97-111 of human IL-21 receptor precursor.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Biochem/physiol Actions

Interleukin-21 (IL-21) is a cytokine that regulates both innate and adaptive immunity. The biological effects of IL-21 are mediated by a heterodimeric receptor made up of IL-21R and a γ chain subunit. IL-21R is expressed in liver, thymus, dendritic cells , lymph nodes and spleen. Binding of IL-21 to the receptor leads to activation of JAK (JAK1 and JAK3), STAT (STAT1, 3, 4 and 5) proteins. IL-21R is important for sustained T cell responses during chronic viral infections, production of interferons against pathogens and protection against tumor formation.

Safety & Documentation

Milli-Q® Water Purification Solutions
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?