• USA Home
  • I6508 - N2-Isobutyryl-5′-O-(4,4′-dimethoxytrityl)-2′-deoxyguanosine 3′-O-succinic acid

EMAIL THIS PAGE TO A FRIEND
I6508 Sigma-Aldrich

N2-Isobutyryl-5′-O-(4,4′-dimethoxytrityl)-2′-deoxyguanosine 3′-O-succinic acid

~95%

Purchase

Properties

Related Categories Biochemicals and Reagents, Nucleoside Analogs, Nucleosides, Nucleotides, Oligonucleotides More...
assay   ~95%
form   solid
storage temp.   −20°C
SMILES string   COc1ccc(cc1)C(OCC2OC(CC2OC(=O)CCC(O)=O)n3cnc4C(=O)N=C(NC(=O)C(C)C)Nc34)(c5ccccc5)c6ccc(OC)cc6
InChI   1S/C39H41N5O10/c1-23(2)36(48)42-38-41-35-34(37(49)43-38)40-22-44(35)31-20-29(54-33(47)19-18-32(45)46)30(53-31)21-52-39(24-8-6-5-7-9-24,25-10-14-27(50-3)15-11-25)26-12-16-28(51-4)17-13-26/h5-17,22-23,29-31H,18-21H2,1-4H3,(H,45,46)(H2,41,42,43,48,49)
InChI key   XQFLSGUZSODAOB-UHFFFAOYSA-N
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?