• USA Home
  • I8028 - Anti-IRAK antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
I8028 Sigma-Aldrich

Anti-IRAK antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym: Anti-Interleukin-1 Receptor-Associated Kinase

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies for Epigenetics and Nuclear Signaling, Antibodies for Gene Regulation,
conjugate   unconjugated
clone   polyclonal
biological source   rabbit
application(s)   immunoprecipitation (IP): 2-4 μg using 0.5 mg of human HeLa or THP-1 cell extracts
  western blot: 0.25 μg/mL using human HeLa or THP-1 cell extracts
species reactivity   human
mol wt   antigen 80 kDa (in non-activated cells)
form   buffered aqueous solution
shipped in   dry ice
storage temp.   −20°C
antibody form   IgG fraction of antiserum
antibody product type   primary antibodies
UniProt accession no.   P51617
Gene Information   human ... IRAK1(3654)

Description

General description

The gene IRAK1 (interleukin 1 receptor associated kinase 1) encodes a member of the IRAK family. The members of this family consist of a conserved N-terminal death domain and a central kinase domain. The gene is mapped to human chromosome Xq28.

Immunogen

synthetic peptide corresponding to amino acids 700-712 of the human IL-1 receptor-associated kinase (IRAK).

Physical form

Solution in phosphate buffered saline, containing 0.02% sodium azide.

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detection methods, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?