• USA Home
  • I8533 - Anti-Interleukin-21 antibody produced in goat

EMAIL THIS PAGE TO A FRIEND
I8533 Sigma-Aldrich

Anti-Interleukin-21 antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym: Anti-IL-21

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, IN-IP, Primary Antibodies
conjugate   unconjugated
clone   polyclonal
biological source   goat
application(s)   capture ELISA: suitable
  neutralization: suitable
  western blot: 0.1-0.2 μg/mL
species reactivity   mouse
form   lyophilized powder
storage temp.   −20°C
antibody form   affinity isolated antibody
antibody product type   primary antibodies
UniProt accession no.   Q9ES17
Gene Information   mouse ... Il21(60505)

Description

General description

Interleukin-21 (IL-21) is a cytokine that regulates both innate and adaptive immunity. It is produced mainly by differentiated CD4+ T cells, natural killer T cells, Th17 and Th1 cells and T follicular helper cells. The biological effects of IL-21 are mediated by a heterodimeric receptor made up of IL-21R and a γ chain subunit. IL-21R is expressed in liver, thymus, dendritic cells , lymph nodes and spleen. Binding of IL-21 to the receptor leads to activation of JAK (JAK1 and JAK3), STAT (STAT1, 3, 4 and 5) proteins. It also activates the MAPK signalling. IL-21 is important for sustained T cell responses during chronic viral infections, production of interferons against pathogens and protection against tumor formation. Deregulation of IL-21 production results in autoimmune disorders including systemic lupus erythematosus, multiple sclerosis and gastrointestinal inflammatory diseases
Anti-Interleukin-21 detects mouse interleukin 21. The antibody shows approximately 5% cross-reactivity with recombinant human IL-21 and less than 1% cross-reactivity with recombinant mouse IL-2.

Immunogen

purified, E. coli-derived, recombinant mouse IL-21.

Application

Anti-Interleukin-21 antibody may be used for ELISA at a working concentration of 0.4 μg/ml. For immunoblotting, the recommended concentration is 0.1-0.2 μg/ml. The antibody is suitable for neutralization reactions.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline, pH 7.4, containing 5% trehalose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detection methods, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?