• USA Home
  • I8639 - Indole-3-propionic acid

EMAIL THIS PAGE TO A FRIEND
I8639 Sigma

Indole-3-propionic acid

plant cell culture tested, crystalline

Synonym: 3-(3-Indolyl)propanoic acid, 3-(3-Indolyl)propionic acid, IPA, NSC 3252, NSC 47831

  • CAS Number 830-96-6

  • Empirical Formula (Hill Notation) C11H11NO2

  • Molecular Weight 189.21

  •  Beilstein Registry Number 147733

  •  EC Number 212-600-1

  •  MDL number MFCD00005660

Purchase

Properties

InChI Key   GOLXRNDWAUTYKT-UHFFFAOYSA-N
form   crystalline
color   light yellow
suitability   plant cell culture tested
storage temp.   −20°C

Description

Biochem/physiol Actions

Studied as an adjunct to improve perfusion after liver transplant.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
NM1329000
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?