EMAIL THIS PAGE TO A FRIEND
I9632 Sigma

Isopropylamine

~99% (GC)

Synonym: 2-Aminopropane

  • CAS Number 75-31-0

  • Linear Formula (CH3)2CHNH2

  • Molecular Weight 59.11

  •  Beilstein Registry Number 605259

  •  EC Number 200-860-9

  •  MDL number MFCD00008082

Purchase

Properties

vapor density   2.04 (vs air)
vapor pressure   9.2 psi ( 20 °C)
InChI Key   JJWLVOIRVHMVIS-UHFFFAOYSA-N
assay   ~99% (GC)
autoignition temp.   755 °F
expl. lim.   10.4 %
refractive index   n20/D 1.374(lit.)
bp   33-34 °C(lit.)
density   0.688 g/mL at 20 °C(lit.)
storage temp.   2-8°C
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN1221 - class 3 - PG 1 - Isopropylamine
WGK Germany 
1
RTECS 
NT8400000
Flash Point(F) 
-.4 °F
Flash Point(C) 
-18 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?