• USA Home
  • I9638 - Inhibin-Like Peptide, human

EMAIL THIS PAGE TO A FRIEND
I9638 Sigma

Inhibin-Like Peptide, human

≥97% (HPLC)

  • CAS Number 92879-83-9

  • Empirical Formula (Hill Notation) C153H241N61O41

  • Molecular Weight 3590.93

  •  MDL number MFCD00148271

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Inhibin Fragments, Miscellaneous Peptides, Peptides and Proteins,
assay   ≥97% (HPLC)
composition   Peptide content, ~65%
storage temp.   −20°C
Gene Information   human ... INHA(3623), INHBA(3624), INHBB(3625)

Description

Amino Acid Sequence

His-Asn-Lys-Gln-Glu-Gly-Arg-Asp-His-Asp-Lys-Ser-Lys-Gly-His-Phe-His-Arg-Val-Val-Ile-His-His-Lys-Gly-Gly-Lys-Ala-His-Arg-Gly

Biochem/physiol Actions

Selectively inhibits the secretion of FSH

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin


Milli-Q® Water Purification Solutions
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?