• USA Home
  • I9908 - Monoclonal Anti-Importin α5/7 antibody produced in rat

EMAIL THIS PAGE TO A FRIEND
I9908 Sigma-Aldrich

Monoclonal Anti-Importin α5/7 antibody produced in rat

clone 2D9, purified immunoglobulin, buffered aqueous solution

Synonym: Anti-Imp α

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Epigenetics and Nuclear Signaling, Antibodies for Gene Regulation, Antibodies to Nuclear Proteins,
conjugate   unconjugated
clone   2D9, monoclonal
biological source   rat
application(s)   immunocytochemistry: suitable
  microarray: suitable
  western blot: 1-2 μg/mL using HeLa cell extract
species reactivity   human, mouse, chicken
mol wt   antigen ~60 kDa
form   buffered aqueous solution
shipped in   dry ice
storage temp.   −20°C
antibody form   purified immunoglobulin
isotype   IgG2a
antibody product type   primary antibodies
UniProt accession no.   P52294
Gene Information   human ... KPNA1(3836), KPNA6(23633)
mouse ... Kpna1(16646), Kpna6(16650)

Description

Immunogen

recombinant mouse importin α5/7.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Safety & Documentation

Safety Information

WGK Germany 
-

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detection methods, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Related Products

related product

Product #

Description

Add to Cart

I9658 Monoclonal Anti-Importin α1 antibody produced in rat, clone 1A6, purified immunoglobulin, buffered aqueous solution
I1784 Anti-Importin α antibody, Mouse monoclonal, clone IM-75, purified from hybridoma cell culture

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?