• USA Home
  • IRA900 - Amberlite® Strongly Basic Anion Exchanger, Chloride form

EMAIL THIS PAGE TO A FRIEND
IRA900 Sigma-Aldrich

Amberlite® Strongly Basic Anion Exchanger, Chloride form

macroreticular

Purchase

Properties

form   macroreticular
particle size   16-50 mesh (wet mesh)
capacity   1.0 meq/mL
  4.2 meq/g

Description

Legal Information

Amberlite is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

216585 Amberlite® IRA-900 chloride form, chloride form

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?