• USA Home
  • K1630 - Keller, et al., (K) Marine Enrichment Basal Salt Mixture

EMAIL THIS PAGE TO A FRIEND
K1630 Sigma

Keller, et al., (K) Marine Enrichment Basal Salt Mixture

powder, plant cell culture tested

Purchase

Properties

form   powder
suitability   plant cell culture tested
Featured Industry   Agriculture
storage temp.   2-8°C

Description

Formula variant

Without silicate. With the macro- and micronutrients as described by Keller, et al. (1987).

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 1479 5.1 / PGIII
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?