• USA Home
  • L0260 - Lithium aluminum hydride

EMAIL THIS PAGE TO A FRIEND
L0260 Sigma

Lithium aluminum hydride

Synonym: LAH, Lithium alanate, Lithium tetrahydroaluminate

Purchase

Properties

InChI Key   OCZDCIYGECBNKL-UHFFFAOYSA-N
mp   125 °C (dec.) (lit.)
storage temp.   2-8°C
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 1410 4.3 / PGI
WGK Germany 
1
RTECS 
BD0100000
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

199877 Lithium aluminum hydride, powder, reagent grade, 95%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?