EMAIL THIS PAGE TO A FRIEND
L0641 Sigma-Aldrich

Leu-Asn

  • CAS Number 14608-81-2

  • Empirical Formula (Hill Notation) C10H19N3O4

  • Molecular Weight 245.28

  •  MDL number MFCD00057937

  •  PubChem Substance ID 24896261

Purchase

Properties

storage temp.   −20°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?