EMAIL THIS PAGE TO A FRIEND
L0650 Sigma

Larcollâ„¢

from larch wood

Synonym: (+)-Arabinogalactan, Polyarabinogalactan

Purchase

Properties

Related Categories Cell Biology, Density Gradient Media and Cell Separation, Hematology and Histology, Polysaccharides
biological source   larch wood
mp   >200 °C (dec.) (lit.)

Description

Application

Prepared specifically for density gradient separations of cells, viruses and organelles.

Biochem/physiol Actions

Dietary fiber that enhances immune system function by stimulating natural killer (NK) cell activity.

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin


Milli-Q® Water Purification Solutions
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?