• USA Home
  • L0919 - Monoclonal Anti-Leukemia Inhibitory Factor antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
L0919 Sigma-Aldrich

Monoclonal Anti-Leukemia Inhibitory Factor antibody produced in mouse

clone 9808, purified immunoglobulin, lyophilized powder

Synonym: Anti-CDF, Anti-Cholinergic differentiation factor, Anti-HILDA, Anti-LIF

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies against Proteins/Bioactives/Markers/Receptors for Stem Cell Biology, Antibodies for Cell Biology, Antibodies to Cytokines and Growth Factors,
conjugate   unconjugated
clone   9808, monoclonal
biological source   mouse
application(s)   flow cytometry: 25 μg/mL
  western blot: 1-2 μg/mL
species reactivity   human
form   lyophilized powder
storage temp.   −20°C
antibody form   purified immunoglobulin
isotype   IgG1
antibody product type   primary antibodies
UniProt accession no.   P15018
Gene Information   human ... LIF(3976)

Description

Immunogen

purified, E. coli-derived recombinant human leukemia inhibitory factor.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline with 5% trehalose.

Biochem/physiol Actions

Leukemia inhibitory factor (LIF) is a pleotrophic glycoprotein that belongs to the IL-6 family of cytokines. It is secreted by many cell types such as, fibroblasts, macrophages, T cells, monocytes, liver cells, osteoblasts and cells of endometrium. A variety of signal transduction pathways are triggered in response to LIF including ERK/MAPK, JAK1, PI3K, STAT3, Jun-c Fos and NF-κB. The various functions of LIF include proliferation of adult human T cells, maintenance of pluripotency of embryonic stem cells and inhibition of leukemic cell differentiation.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1

Milli-Q® Water Purification Solutions
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

L1044 Anti-Leukemia Inhibitory Factor antibody produced in goat, affinity isolated antibody, lyophilized powder
L1169 Anti-Leukemia Inhibitory Factor antibody produced in goat, affinity isolated antibody, lyophilized powder
L0544 Monoclonal Anti-Leukemia Inhibitory Factor antibody produced in rat, clone 139124, purified immunoglobulin, lyophilized powder
L0669 Monoclonal Anti-Leukemia Inhibitory Factor antibody produced in mouse, clone 9824, purified immunoglobulin, lyophilized powder
L9152 Anti-Leukemia Inhibitory Factor antibody produced in goat, IgG fraction of antiserum, lyophilized powder
L9277 Anti-Leukemia Inhibitory Factor antibody produced in goat, IgG fraction of antiserum, lyophilized powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?