• USA Home
  • L1509 - Isolectin A4 (BSI-A4) from Bandeiraea simplicifolia (Griffonia simplicifolia)

EMAIL THIS PAGE TO A FRIEND
L1509 Sigma-Aldrich

Isolectin A4 (BSI-A4) from Bandeiraea simplicifolia (Griffonia simplicifolia)

lyophilized powder

Synonym: Bandeirea simplicifolia agglutinin, BS-I, Lectin from Bandeiraea simplicifolia (Griffonia simplicifolia)

Purchase

Properties

form   lyophilized powder
potency   >32 μg per mL (with human blood group B erythrocytes)
  <16 μg per mL (with human blood group A erythrocytes)
composition   Protein, ~95% E1%/280
storage temp.   −20°C

Description

Analysis Note

Agglutination activity is expressed in μg/mL and is determined from serial dilutions of a 1 mg/mL solution using phosphate buffered saline, pH 6.8, containing, for each lectin, calcium, magnesium, and manganese at different concentrations. This activity is the lowest concentration to agglutinate a 2% suspension of appropriate erythrocytes after 1 hr incubation at 25 °C.

Biochem/physiol Actions

BS-I has a major affinity for terminal α-D-galactosyl residues with a secondary affinity for terminal N-acetyl-α-D-galactosaminyl residues.

Other Notes

BS-I is a tetrameric lectin consisting of two types of subunits designated A and B. There are five BS-I isolectins with different subunit composition: BSI-B4, BSI-AB3, BSI-A2B2, BSI-A3B and BSI-A4. BSI-B4 is blood group B specific and has an exclusive affinity for terminal α-D-galactosyl residues, whereas BSI-A4 has blood group A specificity and has a major affinity for terminal N-acetyl-α-D-galactosaminyl residues.

Quality

Highly purified.

Physical form

Contains sodium citrate

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

L0890 Lectin from Bandeiraea simplicifolia (Griffonia simplicifolia), Isolectin A4 (BSI-A4), FITC conjugate, lyophilized powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?