EMAIL THIS PAGE TO A FRIEND
L1627 Sigma-Aldrich

Lys-Asp

  • CAS Number 20556-18-7

  • Empirical Formula (Hill Notation) C10H19N3O5

  • Molecular Weight 261.27

  •  MDL number MFCD00038214

  •  PubChem Substance ID 24896284

Purchase

Properties

storage temp.   −20°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?