• USA Home
  • L4413 - [D-His(Bzl)6]-LH-RH Fragment 2-9

EMAIL THIS PAGE TO A FRIEND
L4413 Sigma-Aldrich

[D-His(Bzl)6]-LH-RH Fragment 2-9

>99% (HPLC)

  • Empirical Formula (Hill Notation) C61H81N17O10

  • Molecular Weight 1212.40

  •  MDL number MFCD03452954

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, LH-RH, Neuropeptides, Peptides and Proteins,
assay   >99% (HPLC)
storage temp.   −20°C
Gene Information   human ... GNRH1(2796), GNRHR(2798)
mouse ... GNRH1(14714), GNRHR(14715)
rat ... GNRH1(25194), GNRHR(81668)

Description

Amino Acid Sequence

His-Trp-Ser-Tyr-His(Bzl)-Leu-Arg-Pro-NHEt

Biochem/physiol Actions

His-Trp-Ser-Tyr-His(Bzl)-Leu-Arg-Pro-NHEt ([D-His(Bzl)6]-LH-RH Fragment 2-9) is a peptide from GnRH (gonadotropin-releasing hormone) agonist, histerelin. GnRH (Glp-His-Trp-Ser-Tyr-Gly-Leu-Arg-Pro-Gly-NH2), which is also referred as LHRH (luteinizing hormone-releasing hormone) or gonadorelin, is crucial for mammalian reproduction and is released from hypothalamic neurons. It is responsible for the secretion of gonadotropins, luteinizing hormone (LH) and follicle-stimulating hormone (FSH), from the pituitary glands. Mutations in GnRH are associated with normosmic congenital hypogonadotropic hypogonadism. Histerelin is used for the treatment of central precocious puberty in children.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?