• USA Home
  • L4888 - Lectin from Vigna radiata (Mung Bean)

EMAIL THIS PAGE TO A FRIEND
L4888 Sigma-Aldrich

Lectin from Vigna radiata (Mung Bean)

ammonium sulfate suspension

Purchase

Properties

form   ammonium sulfate suspension
potency   <0.1 μg per mL
storage temp.   2-8°C

Description

Unit Definition

One unit will hydrolyze 1.0 μmole of p-nitrophenyl α-D-galactoside to p-nitrophenol and D-galactose per min at pH 7.5 at 25°C.

Physical form

Suspension in 2.5 M (NH4)2SO4 containing 0.05 M Tris, pH 7.0, and stabilizer

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?