• USA Home
  • L7017 - Lectin from Triticum vulgaris (wheat)

EMAIL THIS PAGE TO A FRIEND
L7017 Sigma

Lectin from Triticum vulgaris (wheat)

peroxidase conjugate, lyophilized powder

Synonym: WGA, Wheat germ agglutinin

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Immune Cell Signaling and Blood, Immune System Regulation, Lectins and Lectin Antibodies More...
form   lyophilized powder
potency   <40 μg/mL agglutination activity
peroxidase activity   50-200 units/mg protein
conjugate   peroxidase conjugate
storage temp.   −20°C

Description

Biochem/physiol Actions

WGA is not blood group specific but has an affinity for N-acetyl-β-D-glucosaminyl residues and N-acetyl-β-D-glucosamine oligomers. WGA contains no protein-bound carbohydrate.

Linkage

Same as L3892, but packaged in microcone vials for ease of reconstitution and recovery of microliter volumes.

Packaging

0.5, 1 mg in glass bottle

Package size based on protein content.

Preparation Note

Conjugates are prepared from affinity purified lectin.

Prepared from peroxidase type VI (P 8375) using a modification of the method of O′Sullivan, which favors low molecular weight conjugates. Repurified after conjugation by affinity chromatography.

Unit Definition

One unit will form 1.0 mg of purpurogallin from pyrogallol in 20 sec at pH 6.0 at 20 °C.

Price and Availability


All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?