• USA Home
  • L8649 - Anti-Leukotriene B4 antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
L8649 Sigma-Aldrich

Anti-Leukotriene B4 antibody produced in rabbit

whole antiserum, lyophilized powder

Synonym: Anti-LTB4

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies to Lipids in Cell Signaling, L-LL,
conjugate   unconjugated
clone   polyclonal
biological source   rabbit
application(s)   radioimmunoassay: suitable
species reactivity   human
form   lyophilized powder
storage temp.   2-8°C
antibody form   whole antiserum
antibody product type   primary antibodies
UniProt accession no.   Q15722
Gene Information   human ... LTB4R(1241)

Description

Immunogen

Leukotriene-B4-BSA

Reconstitution

Reconstitute with 5 ml 0.01 M phosphate buffered saline, pH 7.4, containing 0.1% bovine serum albumin and 0.1% sodium azide. Dilute 10-fold in same buffer to obtain working dilution.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Analysis Note

Tests per vial and specificity are determined using a dextran coated charcoal radioimmunoassay (RIA).

Safety & Documentation

Safety Information

WGK Germany 
-

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?