• USA Home
  • L9794 - Anti-Livin α antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
L9794 Sigma-Aldrich

Anti-Livin α antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym: Anti-BIRC7, Anti-Baculoviral IAP repeat-containing 7, Anti-KIAP, Anti-MLIAP, Anti-RNF50

  •  NACRES NA.41

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Apoptosis, Antibodies for Cell Biology, Antibodies to Suppressors of Apoptosis,
conjugate   unconjugated
clone   polyclonal
biological source   rabbit
application(s)   immunoprecipitation (IP): 5-10 μg using lysate of HEK-293T cells expressing human Livin
  western blot: 2.5-5.0 μg/mL using whole extract of HEK-293T cells expressing human Livin
species reactivity   human
mol wt   antigen ~39 kDa
form   buffered aqueous solution
shipped in   dry ice
storage temp.   −20°C
packaging   antibody small pack of 25 μL
antibody form   affinity isolated antibody
Quality Level   200
antibody product type   primary antibodies
concentration   ~1.0 mg/mL
UniProt accession no.   Q96CA5
Gene Information   human ... BIRC7(79444)

Description

General description

Livin or baculoviral IAP repeat containing 7 (BIRC7) gene is mapped to human chromosome 20q13.33 and is expressed in α and β encoding isoforms. Livin-α is an isoform of livin and comprises additional 18 amino acids as compared to Livin-β. This amino acid stretch is located between the BIR and really interesting new gene (RING) domains.

Specificity

Anti-Livin α specifically recognizes human Livin/BIRC7 isoform α.

Application

Anti-Livin α antibody produced in rabbit may be used in immunoblotting and immunoprecipitation.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2–8 °C for up to one month. For extended storage, freeze in working aliquots at –20 °C. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog, our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Biochem/physiol Actions

Livin α protects cells from apoptosis induced by staurosporine. It shows a positive expression bladder tumor.

Safety & Documentation

Safety Information

WGK Germany 
1
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detection methods, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

L9669 Anti-Livin (N-terminal) antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution
L9544 Anti-Livin (C-terminal) antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?