• USA Home
  • LO2218 - Nuclear Receptor Signaling Ligand Set

EMAIL THIS PAGE TO A FRIEND
LO2218 Sigma-Aldrich

Nuclear Receptor Signaling Ligand Set

Purchase

Properties

shipped in   dry ice
storage temp.   −20°C

Description

General description

A set of 55 compounds arranged in 96-well format, one compound per well (250 μL at 10 mM in DMSO).

Features and Benefits

SDFile provided to construct a database containing:
• Structure
• Primary Name
• Secondary Name
• Pharmacological Activity
• Product Number
• Rack Position

The Nuclear Receptor Signaling Ligand Set includes the latest endogenous and synthetic ligands for the following receptors in the field of Nuclear Receptor Signaling:

• Androgen (AR)
• Estrogen (ER)
• Glucocorticoid (GR)
• Mineralocorticoid (MR)
• Peroxisome proliferator-activated alpha (PPARα)
• PPARχ
• PPARδ
• Progesterone (PR)
• Liver X (LXR)
• Retinoic acid (RAR)
• Retinoid X (RXR)
• Thyroid hormone
• Vitamin D

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles

Articles

Retinoic Acid and Gene Expression

All-trans retinoic acid (RA, ATRA) is a pleiotropic activation factor that regulates genes associated with normal vertebrate cellular processes such as cell differentiation, cell proliferation, apopt...
BioFiles 2008, 3.9, 12.
Keywords: Apoptosis, Cancer, Cell proliferation, Cellular processes, Chromatin immunoprecipitation, Condensations, Degradations, Diseases, Gene expression, Hormones, Immunoprecipitation, Ligands, Metabolism, Oxidations, Phosphorylations, Polymerase chain reaction, Sequences, Teratogens, Transcription, Type, Ubiquitinations, Usage, Vitamins

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?