• USA Home
  • M1010 - 4-Methylumbelliferyl stearate

EMAIL THIS PAGE TO A FRIEND
M1010 Sigma-Aldrich

4-Methylumbelliferyl stearate

Purchase

Properties

storage temp.   −20°C
SMILES string   CCCCCCCCCCCCCCCCCC(=O)Oc1ccc2C(C)=CC(=O)Oc2c1
InChI   1S/C28H42O4/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-27(29)31-24-19-20-25-23(2)21-28(30)32-26(25)22-24/h19-22H,3-18H2,1-2H3
InChI key   QOIJAZYHGUNKLA-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?