EMAIL THIS PAGE TO A FRIEND
M313060 Sigma-Aldrich

CUTAG CPG Column

pore size 500 Å, 50 nmol, configured for ABI, configured for PerkinElmer 8900

Purchase

Properties

Related Categories ABI 39X, CUTAG CPG 500A Integral Column for ABI and Expedite, CUTAG Column, Chemical Synthesis, Columns for Synthesizer,
product line   Proligo Reagents
packaging   pkg of 100 ea M313060-100EA
pore size   500 Å
compatibility   configured for ABI
  configured for PerkinElmer 8900
storage temp.   2-8°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?