EMAIL THIS PAGE TO A FRIEND
M3191 Sigma-Aldrich

Myoseverin

≥97% (HPLC), solid

Synonym: 9-Isoproyl-N2,N6-bis-(4-methoxybenxyl)-9H-purine-2,6-diamine

Purchase

Properties

assay   ≥97% (HPLC)
form   solid
color   off-white
solubility   chloroform: soluble 50 mg/mL
shipped in   dry ice
storage temp.   −20°C
SMILES string   COc1ccc(CNc2nc(NCc3ccc(OC)cc3)c4ncn(C(C)C)c4n2)cc1

Description

Biochem/physiol Actions

Myoseverin, a 2,6,9 trisubstituted purine, is a microtuble-binding molecule and reversible inhibitor of tubulin polymerization. It arrests cell cycle in the G2/M phase and causes mature, terminally differentiated mouse myotubes to disassemble into mononucleated fragments capable of responding to growth and differentiation conditions.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?