EMAIL THIS PAGE TO A FRIEND
M4193 Sigma-Aldrich

Maize SSR Primer Set

Purchase

Properties

storage temp.   −20°C

Description

General description

The Maize SSR Primer Set is a cost-effective solution for SSR or SSLP mapping and screening projects that reduces or eliminates the need for custom oligos. The Maize SSR Primer Set consists of 384 pairs of primers selected from the maize genome database. The selections cover the entire maize genome with an average of 20 cM units of map distance between two SSR markers. This primer set is useful for screening new maize populations for genetic mapping, genotyping, marker-trait association analysis, and physical mapping of BAC clones, etc. All primers pass sequence and PCR validation tests. The primers are arrayed in four 96-well plates with a normalized concentration of 50 μM / 200 μl (1 nmol / well). For additional information, please see www.sigma-aldrich.com/maizessr

Packaging

• Four 96-well plates containing 384 pairs of lyophilized primers (1 nmol per well)
• Four additional 96-well plates for dilutions
• 20 sheets of sealing films

Other Notes

The difference between the Maize SSR Primer Set (M4193) and the Maize Polymorphic SSR Primer Set (M8818) is the intention of the sets. The Maize SSR Primer Set is intended to provide relatively even coverage of the entire maize genome (average of 20 cM units of map distance between SSR markers). The Maize Polymorphic SSR Primer Set provides coverage of the entire maize genome, however, it is intented is to target genome regions that have a high probability of polymorphism, with little regard to spacing between SSR markers.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?