• USA Home
  • M4903 - 4-Methylumbelliferyl elaidate

EMAIL THIS PAGE TO A FRIEND
M4903 Sigma-Aldrich

4-Methylumbelliferyl elaidate

95-97% (GC)

Purchase

Properties

assay   95-97% (GC)
storage temp.   −20°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?