• USA Home
  • M6809 - Anti-Matrix Metalloproteinase-24, N-Terminal antibody produced in rabbit

M6809 Sigma

Anti-Matrix Metalloproteinase-24, N-Terminal antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous glycerol solution

Synonym: Anti-MMP-24, Anti-MT5-MMP



Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies to Extracellular Matrix, Antibodies to Matrix Metalloproteinases and TIMPs,
species reactivity   mouse, human
application(s)   flow cytometry: suitable
  immunohistochemistry (frozen sections): suitable
  immunoprecipitation (IP): suitable
  indirect ELISA: suitable
  western blot: 1:2,000
clone   polyclonal
concentration   ~1 mg/mL
antibody form   affinity isolated antibody
form   buffered aqueous glycerol solution
mol wt   antigen mol wt 65 kDa
shipped in   wet ice
storage temp.   −20°C
Gene Information   human ... MMP24(10893)
mouse ... Mmp24(17391)
biological source   rabbit
conjugate   unconjugated



synthetic peptide corresponding to the N-terminal of mouse matrix metalloproteinase-24 (membrane-type matrix metalloproteinase-5).

General description

Matrix metalloproteinase-24 (MMP-24) is expressed majorly in the nervous system and also in the lungs, kidneys and liver. It is a 64kDa protein whose gene is localized on human chromosome 20.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 50% glycerol and 15 mM sodium azide.


Reacts with reduced and native MMP-24. Recognizes the pro-form of MMP-24 as well as activation/breakdown products.

Biochem/physiol Actions

Matrix metalloproteinase-24 (MMP-24) is involved in the conversion of pro-matrix metalloproteinase-2 to its functional active form. It may be important for brain development and studies have shown that it is overexpressed in gastric cancer.

Price and Availability

Explore with Confidence

Expore Enzymes, Proteins & Cofactors

Biomedical Applications
Safety & Documentation

Safety Information

WGK Germany  1
Protocols & Articles


Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy


Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers


Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?