• USA Home
  • M7628 - α-Methyl-DL-tyrosine

EMAIL THIS PAGE TO A FRIEND
M7628 Sigma-Aldrich

α-Methyl-DL-tyrosine

Synonym: DL-2-Methyl-3-(4-hydroxyphenyl)alanine, AMPT

  • CAS Number 658-48-0

  • Linear Formula 4-(HO)C6H4CH2C(CH3)(NH2)CO2H

  • Molecular Weight 195.22

  •  Beilstein/REAXYS Number 2938704

  •  EC Number 211-523-0

  •  MDL number MFCD00004187

Purchase

Properties

InChI Key   NHTGHBARYWONDQ-UHFFFAOYSA-N
mp   >300 °C (lit.)
storage temp.   −20°C

Description

Biochem/physiol Actions

Tyrosine hydroxylase inhibitor.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
YP2890000
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

120693 α-Methyl-DL-tyrosine, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?