EMAIL THIS PAGE TO A FRIEND
M8150 Sigma-Aldrich

Magnesium sulfate

suitable for plant cell culture

Purchase

Properties

vapor density   <0.01 (vs air)
vapor pressure   <0.1 mmHg ( 20 °C)
application(s)   cell culture | plant: suitable
SMILES string   [Mg++].[O-]S([O-])(=O)=O
InChI   1S/Mg.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+2;/p-2
InChI key   CSNNHWWHGAXBCP-UHFFFAOYSA-L
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
OM4500000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?