• USA Home
  • M8444 - Matrix Metalloproteinase-2 from rat fibroblasts

EMAIL THIS PAGE TO A FRIEND
M8444 Sigma-Aldrich

Matrix Metalloproteinase-2 from rat fibroblasts

≥95% (SDS-PAGE), buffered aqueous glycerol solution

Synonym: Collagenase Type IV 72 kD, Gelatinase 72 kD, Gelatinase-A, MMP-2

Purchase

Properties

assay   ≥95% (SDS-PAGE)
form   buffered aqueous glycerol solution
shipped in   dry ice
storage temp.   −70°C
Gene Information   rat ... Mmp2(81686)

Description

Biochem/physiol Actions

Degrades general matrix components and may have a role in processes such as host defense, cell proliferation, and protein turnover as well as tissue remodeling.

Quality

Essentially free of other matrix metalloproteinases and tissue inhibitors of metalloproteinases (TIMPS).

Physical form

Solution in 50% glycerol (v/v) containing 10 mM sodium phosphate, pH 7.4, and 75 mM sodium chloride.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 1
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?