EMAIL THIS PAGE TO A FRIEND
M8905 Sigma-Aldrich

5-Methyluridine

powder

Synonym: Ribothymidine

Purchase

Properties

Related Categories Biochemicals and Reagents, Chemical Biology, Chemical Synthesis, Nucleobase, Nucleoside and Nucleotide Derivatives, Nucleoside Analogs,
assay   ≥99% (HPLC)
  ≥99% (TLC)
form   powder
color   white
mp   183-184 °C (lit.)
solubility   water: 50 mg/mL, clear, colorless
storage temp.   2-8°C
SMILES string   CC1=CN([C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O)C(=O)NC1=O
InChI   1S/C10H14N2O6/c1-4-2-12(10(17)11-8(4)16)9-7(15)6(14)5(3-13)18-9/h2,5-7,9,13-15H,3H2,1H3,(H,11,16,17)/t5-,6-,7-,9-/m1/s1
InChI key   DWRXFEITVBNRMK-JXOAFFINSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

HPLC Analysis of Nucleosides on Ascentis® Express OH5 (HILIC Mode)

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Nucleosides on Ascentis® Express OH5 (HILIC Mode)
Keywords: Antivirals, Chromatography, High performance liquid chromatography, Metabolism

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?