• USA Home
  • M8940 - Methyl 14(S),15(S)-epoxy-13(R)-hydroxy-(5Z,8Z,11Z)-eicosatrienoate

EMAIL THIS PAGE TO A FRIEND
M8940 Sigma-Aldrich

Methyl 14(S),15(S)-epoxy-13(R)-hydroxy-(5Z,8Z,11Z)-eicosatrienoate

≥95% (HPLC), hexane solution

  • Empirical Formula (Hill Notation) C21H34O4

  • Molecular Weight 350.49

  •  EC Number 203-777-6

  •  MDL number MFCD04040010

  •  PubChem Substance ID 329818074

  •  NACRES NA.25

Purchase

Properties

Related Categories Arachidonic Acid Cascade, Cell Biology, Cell Signaling and Neuroscience, Cyclooxygenase, Enzyme Inhibitors,
Quality Level   200
assay   ≥95% (HPLC)
form   hexane solution
functional group   ester
shipped in   dry ice
storage temp.   −20°C
SMILES string   CCCCCC1OC1[C@H](O)\C=C/C\C=C/C\C=C/CCCC(=O)OC
InChI   1S/C21H34O4/c1-3-4-12-16-19-21(25-19)18(22)15-13-10-8-6-5-7-9-11-14-17-20(23)24-2/h6-9,13,15,18-19,21-22H,3-5,10-12,14,16-17H2,1-2H3/b8-6-,9-7-,15-13-/t18-,19+,21-/m1/s1
InChI key   DBXPKRNKMVEUMA-LLNXGUOUSA-N

Description

Other Notes

15-Lipoxygenase product of arachidonic acid, formed by a similar biosynthesis as hepoxilins

Safety & Documentation

Safety Information

Signal word 
Danger
Target organs 
Central nervous system, Nervous system
RIDADR 
UN1208 - class 3 - PG 2 - Hexanes
WGK Germany 
WGK 2
Flash Point(F) 
-14.8 °F - closed cup - Not applicable
Flash Point(C) 
-26 °C - closed cup - Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?