EMAIL THIS PAGE TO A FRIEND
M9036 Sigma-Aldrich

Metronidazole

analytical standard

Synonym: 2-Methyl-5-nitroimidazole-1-ethanol

  • CAS Number 443-48-1

  • Empirical Formula (Hill Notation) C6H9N3O3

  • Molecular Weight 171.15

  •  Beilstein/REAXYS Number 611683

  •  EC Number 207-136-1

  •  MDL number MFCD00009750

  •  PubChem Substance ID 24897314

Purchase

Properties

grade   analytical standard
packaging   vial of 200 mg
mp   159-161 °C (lit.)
Mode of action   DNA synthesis | interferes
storage temp.   2-8°C
SMILES string   CC1=NC=C([N+]([O-])=O)N1CCO
InChI   1S/C6H9N3O3/c1-5-7-4-6(9(11)12)8(5)2-3-10/h4,10H,2-3H2,1H3
InChI key   VAOCPAMSLUNLGC-UHFFFAOYSA-N

Description

General description

Chemical structure: imidazole

Packaging

Supplied in amber screw-cap vials

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
RTECS 
NI5600000
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?