• USA Home
  • M9279 - N,N′-Diisopropyl-O-methylisourea

EMAIL THIS PAGE TO A FRIEND
M9279 Sigma

N,N′-Diisopropyl-O-methylisourea

≥97% (GC)

Synonym: O-Methyl-N,N′-diisopropylisourea

  • CAS Number 54648-79-2

  • Linear Formula (CH3)2CHNHC(OCH3)=NCH(CH3)2

  • Molecular Weight 158.24

  •  Beilstein Registry Number 1753052

  •  EC Number 259-275-2

  •  MDL number MFCD00008866

Purchase

Properties

InChI Key   PUVRRPLSJKDMKH-UHFFFAOYSA-N
assay   ≥97% (GC)
density   0.872 g/mL at 20 °C(lit.)
storage temp.   2-8°C
Safety & Documentation

Safety Information

RIDADR 
UN 1993 3/PG 3
WGK Germany 
3
Flash Point(F) 
95 °F
Flash Point(C) 
35 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

226408 o-Methyl-N,N′-diisopropylisourea, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?