• USA Home
  • MAK375 - Covidyte TF670 Substrate
EMAIL THIS PAGE TO A FRIEND
MAK375 Sigma-Aldrich
Covidyte TF670 Substrate
New
sufficient for 100 fluorometric tests
Purchase
Description
General description
The coronavirus main protease, which plays a pivotal role in viral gene expression and replication through the proteolytic processing of replicase polyproteins, is an attractive target for anti-CoV drug design. The inhibition of viral proteases necessary for proteolytic processing of polyproteins has been a successful strategy in the treatment of human immunodeficiency virus (HIV) and hepatitis C respectively, proving the potential of protease inhibitors for the treatment of viral infections. Similarly, the main protease of SARS-CoV-2 is thought to be essential for viral replication and is therefore regarded as a promising target for antiviral therapy of Covid-19.
Features and Benefits
Compatible with high-throughput handling systems.
Suitability
Suitable for developing methods to screen potential inhibitors of coronavirus proteases
Principle
Covidyte TF670 is a peptide substrate containing a 14-amino acid sequence that can be cleaved by coronavirus proteases. The dark-FRET peptide contains Tide Quencher 5 (TQ5) as a quencher and Tide Fluor 5 (TF5) as a fluorescent donor on the N- and C-terminals respectively where the fluorescence of TF5 is effectively quenched by TQ5 when the peptide is intact. When the peptide is hydrolyzed by coronavirus proteases, the TF5 fragment generates significantly enhanced fluorescence since its fluorescence is no longer quenched by TQ5. The activity of coronavirus proteases can be effectively monitored by the fluorescence intensity of TF5.
Legal Information
Covidyte is a trademark of AAT Bioquest
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis (COA)
Please Enter a Lot Number
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?