EMAIL THIS PAGE TO A FRIEND
MYC1 Sigma

Mycoplasma Stain Kit

Purchase

Description

Application

The Mycoplasma Stain Kit is designed for in situ detection of mycoplasma and other prokaryotic organisms in cell cultures. This procedure is based on the Hoechst Stain method cited by the Tissue Culture Association (TCA Procedure No. 75361).

Price and Availability


All labs need water

Standard component

Description

Product #

Add to Cart

Hanks' Balanced Salt Solution without phenol red and sodium bicarbonate 3 x 35 mL SDS    
Hoechst Stain Solution 10 mL SDS    
Mounting Medium 10 mL SDS    
negative control slides 10 SDS    
positive control slides 10 SDS    
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
Protocols & Articles

Articles

Mycoplasma Detection/Elimination

The maintenance of contamination-free cell lines is essential to cell-based research. Among the biggest concerns are mycoplasma contamination. Although mycoplasma do not usually kill contaminated cel...
Don Finley
BioFiles v7 n1, 2011, 14–15
Keywords: Eliminations, Metabolism, Polymerase chain reaction

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?