• USA Home
  • N0274 - Anti-Neurturin antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
N0274 Sigma-Aldrich

Anti-Neurturin antibody produced in rabbit

~0.5 mg/mL, affinity isolated antibody

Synonym: Anti-NRTN, Anti-NTN

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies for Kinase/Phosphatase Biology, Antibodies to Cytokines and Growth Factors,
conjugate   unconjugated
clone   polyclonal
biological source   rabbit
application(s)   western blot: 1:500 using a human HeLa cell lysate
species reactivity   human
mol wt   antigen 14 kDa
shipped in   dry ice
storage temp.   −20°C
antibody form   affinity isolated antibody
antibody product type   primary antibodies
concentration   ~0.5 mg/mL
UniProt accession no.   Q99748
Gene Information   human ... NRTN(4902)

Description

General description

Neurturin (NTN) belongs to a novel class of glial-derived neurotrophic factors that have neuroprotective and neuroregenerative effects. NTN promotes the survival of neurons in central and peripheral nervous by signaling via the receptor GFRα2 and a transmembrane tyrosine kinase, Ret. The pathways through which NTN produces effects on parasympathetic neurons are MAPK, PI3K and CREB-mediated signaling. NTN plays a major role in the development and maintenance of postganglionic parasympathetic neurons. The functions of NTN include regulation of spermatogenesis, kidney morphogenesis and control of smooth muscle of penile blood vessels and several intrapelvic organs, especially urogenital organs. Anti-Neurturin recognizes human neurturin.

Immunogen

synthetic peptide corresponding to amino acids 178-193 of human neurturin.

Application

Anti-Neurturin antibody may be used for immunoblotting at a working antibody dilution of 1:500 using whole HeLa cell lysate.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Safety & Documentation

Safety Information

WGK Germany 
-

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detection methods, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?