• USA Home
  • N0502 - 4-Nitrophenyl hexanoate

EMAIL THIS PAGE TO A FRIEND
N0502 Sigma-Aldrich

4-Nitrophenyl hexanoate

liquid

Synonym: p-Nitrophenyl caproate, Caproic acid p-nitrophenyl ester

  • CAS Number 956-75-2

  • Empirical Formula (Hill Notation) C12H15NO4

  • Molecular Weight 237.25

  •  Beilstein/REAXYS Number 5033817

  •  MDL number MFCD00056111

Purchase

Properties

form   liquid
color   light yellow
storage temp.   −20°C
SMILES string   CCCCCC(=O)Oc1ccc(cc1)[N+]([O-])=O
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?