• USA Home
  • N0540 - Nicotinic Receptor, Neuronal human

EMAIL THIS PAGE TO A FRIEND
N0540 Sigma-Aldrich

Nicotinic Receptor, Neuronal human

membrane suspension

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, G Protein Function, G Proteins and Cyclic Nucleotides, G-Protein Coupled Receptors (GPCR),
biological source   (neuroblastoma cell membranes)
form   membrane suspension
concentration   ~8.3 mg/mL protein
shipped in   dry ice
storage temp.   −70°C
Gene Information   human ... CHRNA2(1135), CHRNB2(1141)

Description

General description

α-bungarotoxin insensitive

Biochem/physiol Actions

Membranes are suspended in 50 mM Tris (pH 7.4 at 25 °C).

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Milli-Q® Water Purification Solutions
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?