EMAIL THIS PAGE TO A FRIEND
N0638 Sigma-Aldrich

New Fuchsin

Dye content ~80 %

Synonym: Basic Violet 2, Fuchsin N, Magenta III

  • CAS Number 3248-91-7

  • Linear Formula C22H24N3Cl

  • Molecular Weight 365.90

  •  Colour Index Number 42520

  •  Beilstein/REAXYS Number 8740867

  •  EC Number 221-831-7

  •  MDL number MFCD00012567

Purchase

Properties

composition   Dye content, ~80%
λmax   553 nm
SMILES string   [Cl-].Cc1cc(ccc1N)C(\c2ccc(N)c(C)c2)=C3/C=CC(=[NH2+])C(C)=C3
InChI   1S/C22H23N3.ClH/c1-13-10-16(4-7-19(13)23)22(17-5-8-20(24)14(2)11-17)18-6-9-21(25)15(3)12-18;/h4-12,23H,24-25H2,1-3H3;1H
InChI key   IPSIPYMEZZPCPY-UHFFFAOYSA-N

Description

Legal Information

Magenta is a trademark of Magenta Corp.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

72200 New Fuchsin, for microscopy

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?