EMAIL THIS PAGE TO A FRIEND
N0641 SAFC

Nisoldipine

~97%

Synonym: (±)-Isobutyl methyl 1,4-dihydro-2,6-dimethyl-4-(o-nitrophenyl)-3,5-pyridinedicarboxylate, 1,4-Dihydro-2,6-dimethyl-4-(2-nitrophenyl)-3,5-pyridinedicarboxylic acid methyl 2-methylpropyl ester, Sular

Purchase

Properties

assay   ~97%
storage temp.   2-8°C
SMILES string   COC(=O)C1=C(C)NC(C)=C(C1c2ccccc2[N+]([O-])=O)C(=O)OCC(C)C
InChI   1S/C20H24N2O6/c1-11(2)10-28-20(24)17-13(4)21-12(3)16(19(23)27-5)18(17)14-8-6-7-9-15(14)22(25)26/h6-9,11,18,21H,10H2,1-5H3
InChI key   VKQFCGNPDRICFG-UHFFFAOYSA-N
Gene Information   human ... CACNA1C(775), CACNA1D(776), CACNA1F(778), CACNA1S(779)

Description

Biochem/physiol Actions

L-type (CaV1.2) calcium channel blocker; dihydropyridine-type calcium channel blocker with selectivity for smooth muscle (CaV1.2b) over cardiac muscle (CaV1.2a). Arterial vasodilator and antihypertensive agent.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
US7975600

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?