• USA Home
  • N0899 - Anti-Neuropilin-1 antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
N0899 Sigma

Anti-Neuropilin-1 antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Purchase

Properties

Related Categories Alphabetical Index, Angiogenesis, Antibodies, Antibodies for Neurobiology, Antibodies to Neural Proteins,
species reactivity   rat, mouse
application(s)   immunohistochemistry: suitable
  western blot: suitable
clone   polyclonal
antibody form   IgG fraction of antiserum
form   buffered aqueous solution
shipped in   wet ice
storage temp.   2-8°C
Gene Information   human ... NRP1(8829)
mouse ... Nrp1(18186)
rat ... Nrp1(246331)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

DLDKKNTEIKIDETG, highly purified peptide corresponding to amino acids 813-827 of rat neuropilin-1 conjugated to KLH. Identical in rat and mouse. Highly homologous (14/15 residues identical) in human.

Physical form

Solution in 0.05 M sodium phosphate buffer containing 0.1% sodium azide and 0.2% gelatin.

Price and Availability


Antibody Explorer

Explore with Confidence

All labs need water
Safety & Documentation

Safety Information

WGK Germany  2
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

N3287 Monoclonal Anti-Neuropilin-1 antibody produced in mouse, clone 130603, purified immunoglobulin, lyophilized powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?