• USA Home
  • N3159 - N-(NBD-Aminolauroyl)sphingomyelin

EMAIL THIS PAGE TO A FRIEND
N3159 Sigma-Aldrich

N-(NBD-Aminolauroyl)sphingomyelin

>98%

  • Empirical Formula (Hill Notation) C41H73N6O9P

  • Molecular Weight 825.03

  •  MDL number MFCD02259246

  •  PubChem Substance ID 329818603

Purchase

Properties

Related Categories Alphabetical, Biochemicals and Reagents, Cell Biology, Cell Signaling and Neuroscience, Complex Lipids,
assay   >98%
shipped in   dry ice
storage temp.   −20°C
SMILES string   CCCCCCCCCCCCC\C=C\[C@@H](O)[C@H](COP([O-])(=O)OCC[N+](C)(C)C)NC(=O)CCCCCCCCCCCNc1ccc([N+]([O-])=O)c2nonc12
InChI   1S/C41H73N6O9P/c1-5-6-7-8-9-10-11-12-13-15-18-21-24-27-38(48)36(34-55-57(52,53)54-33-32-47(2,3)4)43-39(49)28-25-22-19-16-14-17-20-23-26-31-42-35-29-30-37(46(50)51)41-40(35)44-56-45-41/h24,27,29-30,36,38,48H,5-23,25-26,28,31-34H2,1-4H3,(H2-,42,43,45,49,52,53)/b27-24+/t36-,38+/m0/s1
InChI key   FCQMKLRMKNYONQ-YPGNEAGUSA-N

Description

General description

Fluorescently labeled fatty acid sphingolipid membrane probe. NBD group excitation 460 nm, emission 534 nm.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?