EMAIL THIS PAGE TO A FRIEND
N3395 Sigma-Aldrich

Nomilin

  • CAS Number 1063-77-0

  • Empirical Formula (Hill Notation) C28H34O9

  • Molecular Weight 514.56

  •  MDL number MFCD00075979

  •  PubChem Substance ID 24897643

Purchase

Properties

SMILES string   CC(=O)OC1CC(=O)OC(C)(C)[C@@H]2CC(=O)[C@]3(C)[C@H](CC[C@@]4(C)[C@@H](OC(=O)[C@H]5O[C@@]345)c6ccoc6)[C@@]12C
InChI   1S/C28H34O9/c1-14(29)34-19-12-20(31)36-24(2,3)17-11-18(30)27(6)16(26(17,19)5)7-9-25(4)21(15-8-10-33-13-15)35-23(32)22-28(25,27)37-22/h8,10,13,16-17,19,21-22H,7,9,11-12H2,1-6H3/t16-,17+,19?,21+,22-,25+,26-,27+,28-/m1/s1
InChI key   KPDOJFFZKAUIOE-BIYZDVJRSA-N

Description

Biochem/physiol Actions

Has anti-proliferative activity on cultured human breast cancer cells.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?