• USA Home
  • N4784 - NF449 octasodium salt

EMAIL THIS PAGE TO A FRIEND
N4784 Sigma-Aldrich

NF449 octasodium salt

solid

Synonym: 4,4′,4″,4″′-[Carbonylbis[imino-5,1,3-benzenetriyl bis(carbonyl-imino)]]tetrakis(benzene-1,3-disulfonic acid) octasodium salt

  • Empirical Formula (Hill Notation) C41H24N6Na8O29S8

  • Molecular Weight 1505.09

  •  MDL number MFCD02179205

Purchase

Properties

form   solid
color   white
solubility   H2O: soluble
storage temp.   2-8°C
SMILES string   [Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[O-]S(=O)(=O)c1ccc(NC(=O)c2cc(NC(=O)Nc3cc(cc(c3)C(=O)Nc4ccc(cc4S([O-])(=O)=O)S([O-])(=O)=O)C(=O)Nc5ccc(cc5S([O-])(=O)=O)S([O-])(=O)=O)cc(c2)C(=O)Nc6ccc(cc6S([O-])(=O)=O)S([O-])(=O)=O)c(c1)S([O-])(=O)=O
Gene Information   human ... P2RX1(5023)

Description

Biochem/physiol Actions

Potent G-protein antagonist, selective for Gαs; highly potent, selective P2X1 receptor antagonist.

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?